Assay Details
| Gene Symbol: YIPF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257440.1 | NP_001244369.1 | 837 | 69 | ||
| GenBank mRNA | JU336692.1 | - | 837 | 69 | ||
| JU477486.1 | - | 830 | 69 | |||
| JV048434.1 | - | 833 | 69 | |||
More Information
Set Membership: |
|

YIPF4
Genomic Map