Assay Details
Gene Symbol: KIRREL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001116954.4 | XP_001116954.2 | 1871 | 60 | ||
XM_015112606.2 | XP_014968092.2 | 1919 | 60 | |||
GenBank mRNA | JV043768.1 | - | 1812 | 60 | ||
JV635438.1 | - | 1817 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
