Assay Details
Gene Symbol: HSD17B11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257582.1 | NP_001244511.1 | 405 | 63 | ||
XM_015138742.2 | XP_014994228.2 | 692 | 63 | |||
GenBank mRNA | JU334817.1 | - | 405 | 63 | ||
JU475845.1 | - | 325 | 63 | |||
JV047260.1 | - | 303 | 63 | |||
JV636197.1 | - | 355 | 63 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
