Assay Details
| Gene Symbol: FBXW4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257946.2 | NP_001244875.1 | 967 | 118 | ||
| XM_015147984.2 | XP_015003470.2 | 1227 | 118 | |||
| XM_015147985.2 | XP_015003471.2 | 1227 | 118 | |||
| XM_028853483.1 | XP_028709316.1 | 513 | 118 | |||
| XM_028853484.1 | XP_028709317.1 | 434 | 118 | |||
| GenBank mRNA | JU335934.1 | - | 967 | 118 | ||
| JU476815.1 | - | 1002 | 118 | |||
| JU476816.1 | - | 1002 | 118 | |||
| JV047943.1 | - | 1002 | 118 | |||
| JV721232.1 | - | 634 | 118 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

FBXW4
Genomic Map