Assay Details
| Gene Symbol: SPAG7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015118374.2 | XP_014973860.2 | 268 | 116 | ||
| GenBank mRNA | JV528665.1 | - | 108 | 116 | ||
| JV669736.1 | - | 82 | 116 | |||
More Information
Set Membership: |
|

SPAG7
Genomic Map