Assay Details
| Gene Symbol: LOC711104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001108444.4 | XP_001108444.1 | 998 | 68 | ||
| XM_015135024.2 | XP_014990510.1 | 671 | 68 | |||
| XM_015135025.2 | XP_014990511.1 | 1003 | 68 | |||
| XM_015135026.2 | XP_014990512.1 | 3353 | 68 | |||
| XM_015135028.2 | XP_014990514.1 | 2873 | 68 | |||
| XM_015135034.2 | XP_014990520.1 | 932 | 68 | |||
| XM_015135036.2 | XP_014990522.1 | 4457 | 68 | |||
| XM_015135040.2 | XP_014990526.1 | 1068 | 68 | |||
| XM_028846365.1 | XP_028702198.1 | 1687 | 68 | |||
| GenBank mRNA | JU319762.1 | - | 885 | 68 | ||
| JU471676.1 | - | 646 | 68 | |||
| JV044892.1 | - | 804 | 68 | |||
More Information
Set Membership: |
|

LOC711104
Genomic Map