Assay Details
| Gene Symbol: ATP1B3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261524.1 | NP_001248453.1 | 680 | 106 | ||
| XM_028843417.1 | XP_028699250.1 | 779 | 106 | |||
| XM_028843418.1 | XP_028699251.1 | 694 | 106 | |||
| XM_028843419.1 | XP_028699252.1 | 780 | 106 | |||
| GenBank mRNA | JU334677.1 | - | 683 | 106 | ||
| JU475733.1 | - | 608 | 106 | |||
| JV047170.1 | - | 680 | 106 | |||
| JV652485.1 | - | 712 | 106 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ATP1B3
Genomic Map