Assay Details
Gene Symbol: DARS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015110010.2 | XP_014965496.1 | 777 | 96 | ||
XM_028830668.1 | XP_028686501.1 | 607 | 96 | |||
GenBank mRNA | JU333178.1 | - | 711 | 96 | ||
JU474488.1 | - | 723 | 96 | |||
JV046305.1 | - | 637 | 96 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
