Assay Details
| Gene Symbol: CAB39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267016.1 | NP_001253945.1 | 805 | 153 | ||
| XM_015111248.2 | XP_014966734.1 | 864 | 153 | |||
| GenBank mRNA | JU319865.1 | - | 802 | 153 | ||
| JU475973.1 | - | 781 | 153 | |||
| JV045169.1 | - | 805 | 153 | |||
| JV601991.1 | - | 1005 | 153 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CAB39
Genomic Map