Assay Details
| Gene Symbol: RXRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001108780.4 | XP_001108780.3 | 1037 | 70 | ||
| XM_015136101.2 | XP_014991587.2 | 1037 | 70 | |||
| GenBank mRNA | JU328335.1 | - | 660 | 70 | ||
| JV720722.1 | - | 1002 | 70 | |||
More Information
Set Membership: |
|

RXRB
Genomic Map