Assay Details
| Gene Symbol: GABRA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028841928.1 | XP_028697761.1 | 497 | 77 | ||
| XM_028841929.1 | XP_028697762.1 | 440 | 77 | |||
| XM_028841930.1 | XP_028697763.1 | 317 | 77 | |||
| GenBank mRNA | JU333818.1 | - | 429 | 77 | ||
| JV567028.1 | - | 486 | 77 | |||
| JV611896.1 | - | 486 | 77 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028841930.1 transcripts.
Set Membership: |
|

GABRA3
Genomic Map