Assay Details
Gene Symbol: MBD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001082249.4 | XP_001082249.2 | 733 | 62 | ||
XM_015121998.1 | XP_014977484.1 | 772 | 62 | |||
XM_015121999.2 | XP_014977485.1 | 772 | 62 | |||
GenBank mRNA | JU327338.1 | - | 137 | 62 | ||
JV726184.1 | - | 154 | 62 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
