Assay Details
| Gene Symbol: RIPOR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126402.2 | XP_014981888.1 | 1115 | 58 | ||
| XM_015126404.2 | XP_014981890.1 | 1093 | 58 | |||
| XM_015126405.1 | XP_014981891.1 | 1167 | 58 | |||
| XM_028841362.1 | XP_028697195.1 | 1540 | 58 | |||
| XM_028841364.1 | XP_028697197.1 | 1109 | 58 | |||
| GenBank mRNA | JV557734.1 | - | 1070 | 58 | ||
| JV628622.1 | - | 1070 | 58 | |||
More Information
Set Membership: |
|

RIPOR1
Genomic Map