Assay Details
| Gene Symbol: ASPHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002808294.3 | XP_002808340.2 | 1577 | 64 | ||
| GenBank mRNA | JU325967.1 | - | 683 | 64 | ||
| JV577193.1 | - | 1124 | 64 | |||
| JV611823.1 | - | 838 | 64 | |||
More Information
Set Membership: |
|

ASPHD1
Genomic Map