Assay Details
Gene Symbol: LOC704480 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015147233.2 | XP_015002719.2 | 933 | 58 | ||
XM_028826445.1 | XP_028682278.1 | 698 | 58 | |||
XM_028826446.1 | XP_028682279.1 | 716 | 58 | |||
XM_028826447.1 | XP_028682280.1 | 536 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
