Assay Details
| Gene Symbol: SORD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260778.1 | NP_001247707.1 | 867 | 71 | ||
| GenBank mRNA | JU323287.1 | - | 940 | 71 | ||
| JU476912.1 | - | 867 | 71 | |||
More Information
Set Membership: |
|

SORD
Genomic Map