Assay Details
Gene Symbol: PAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001091544.4 | XP_001091544.2 | 979 | 73 | ||
XM_028852756.1 | XP_028708589.1 | 1144 | 73 | |||
XM_028852757.1 | XP_028708590.1 | 1240 | 73 | |||
XM_028852758.1 | XP_028708591.1 | 1399 | 73 | |||
GenBank mRNA | JU470895.1 | - | 975 | 73 | ||
JU470896.1 | - | 964 | 73 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
