Assay Details
Gene Symbol: CYTIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015110187.2 | XP_014965673.2 | 531 | 75 | ||
XM_028830724.1 | XP_028686557.1 | 717 | 75 | |||
GenBank mRNA | JU477946.1 | - | 463 | 75 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
