Assay Details
| Gene Symbol: GUCA1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001102050.4 | XP_001102050.1 | 1224 | 110 | ||
| XM_028844636.1 | XP_028700469.1 | 1224 | 110 | |||
More Information
Set Membership: |
|

GUCA1C
Genomic Map