Assay Details
| Gene Symbol: UBE2Z | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015119345.2 | XP_014974831.2 | 782 | 93 | ||
| GenBank mRNA | JU327376.1 | - | 541 | 93 | ||
| JV659249.1 | - | 513 | 93 | |||
| JV667270.1 | - | 802 | 93 | |||
More Information
Set Membership: |
|

UBE2Z
Genomic Map