Assay Details
| Gene Symbol: PDZRN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001098466.4 | XP_001098466.2 | 1422 | 113 | ||
| XM_015130386.2 | XP_014985872.1 | 762 | 113 | |||
| XM_015130387.2 | XP_014985873.1 | 770 | 113 | |||
| XM_015130388.2 | XP_014985874.1 | 696 | 113 | |||
| XM_028844367.1 | XP_028700200.1 | 1335 | 113 | |||
| GenBank mRNA | JV575240.1 | - | 358 | 113 | ||
More Information
Set Membership: |
|

PDZRN3
Genomic Map