Assay Details
| Gene Symbol: UBE2E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015132279.2 | XP_014987765.1 | 415 | 101 | ||
| XM_028843645.1 | XP_028699478.1 | 709 | 101 | |||
| GenBank mRNA | JU320397.1 | - | 534 | 101 | ||
| JU477053.1 | - | 659 | 101 | |||
| JV048141.1 | - | 658 | 101 | |||
| JV627695.1 | - | 661 | 101 | |||
| JV660450.1 | - | 622 | 101 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

UBE2E1
Genomic Map