Assay Details
Gene Symbol: URGCP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002803295.3 | XP_002803341.1 | 426 | 87 | ||
XM_002803296.3 | XP_002803342.1 | 399 | 87 | |||
XM_015133647.2 | XP_014989133.1 | 1200 | 87 | |||
XM_015133648.2 | XP_014989134.1 | 1173 | 87 | |||
XM_015133649.2 | XP_014989135.1 | 1195 | 87 | |||
XM_015133650.2 | XP_014989136.1 | 270 | 87 | |||
GenBank mRNA | JU326433.1 | - | 148 | 87 | ||
JV049002.1 | - | 393 | 87 | |||
JV667426.1 | - | 152 | 87 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
