Assay Details
| Gene Symbol: KSR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015152906.2 | XP_015008392.1 | 2686 | 59 | ||
| XM_028829953.1 | XP_028685786.1 | 2440 | 59 | |||
| XM_028829954.1 | XP_028685787.1 | 2440 | 59 | |||
| XM_028829955.1 | XP_028685788.1 | 2434 | 59 | |||
More Information
Set Membership: |
|

KSR2
Genomic Map