Assay Details
| Gene Symbol: UQCC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002798160.3 | XP_002798206.2 | 574 | 97 | ||
| XM_028828352.1 | XP_028684185.1 | 393 | 97 | |||
| XM_028828353.1 | XP_028684186.1 | 397 | 97 | |||
| XM_028828354.1 | XP_028684187.1 | 378 | 97 | |||
| GenBank mRNA | JU335284.1 | - | 421 | 97 | ||
| JU476239.1 | - | 412 | 97 | |||
| JV549829.1 | - | 456 | 97 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

UQCC1
Genomic Map