Assay Details
Gene Symbol: PAPSS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266008.1 | NP_001252937.1 | 1280 | 69 | ||
XM_015147608.2 | XP_015003094.2 | 1539 | 69 | |||
GenBank mRNA | JV044612.1 | - | 1285 | 69 | ||
JV044613.1 | - | 1285 | 69 | |||
JV635780.1 | - | 1280 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
