Assay Details
| Gene Symbol: PI4K2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015147870.2 | XP_015003356.1 | 1330 | 65 | ||
| GenBank mRNA | JU327506.1 | - | 1147 | 65 | ||
| JV621375.1 | - | 1161 | 65 | |||
| JV655423.1 | - | 1160 | 65 | |||
More Information
Set Membership: |
|

PI4K2A
Genomic Map