Assay Details
Gene Symbol: MAP2K6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261706.1 | NP_001248635.1 | 1201 | 80 | ||
XM_028835466.1 | XP_028691299.1 | 1225 | 80 | |||
XM_028835467.1 | XP_028691300.1 | 1243 | 80 | |||
GenBank mRNA | JU320442.1 | - | 1208 | 80 | ||
JU472320.1 | - | 1201 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
