Assay Details
| Gene Symbol: MGST2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261313.1 | NP_001248242.1 | 187 | 135 | ||
| XM_015139221.2 | XP_014994707.1 | 320 | 135 | |||
| XM_028847996.1 | XP_028703829.1 | 320 | 135 | |||
| XM_028847998.1 | XP_028703831.1 | 320 | 135 | |||
| GenBank mRNA | JU322009.1 | - | 268 | 135 | ||
| JU473802.1 | - | 307 | 135 | |||
| JV047100.1 | - | 187 | 135 | |||
| KC734514.1 | - | 76 | 135 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MGST2
Genomic Map