Assay Details
| Gene Symbol: EFCAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194172.3 | NP_001181101.1 | 633 | 105 | ||
| XM_015145313.2 | XP_015000799.1 | 633 | 105 | |||
More Information
Set Membership: |
|

EFCAB1
Genomic Map