Assay Details
Gene Symbol: SLC6A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266791.2 | NP_001253720.1 | 852 | 86 | ||
XM_015141181.2 | XP_014996667.1 | 2428 | 86 | |||
XM_015141184.2 | XP_014996670.1 | 653 | 86 | |||
GenBank mRNA | JU335991.1 | - | 86 | |||
JV600973.1 | - | 894 | 86 | |||
JV667283.1 | - | 852 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
