Assay Details
| Gene Symbol: CRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001110878.4 | XP_001110878.2 | 3617 | 76 | ||
| XM_001110912.4 | XP_001110912.2 | 3953 | 76 | |||
| XM_015120817.2 | XP_014976303.1 | 4088 | 76 | |||
| XM_028851727.1 | XP_028707560.1 | 3953 | 76 | |||
| GenBank mRNA | JU333687.1 | - | 3884 | 76 | ||
| JV710805.1 | - | 3903 | 76 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CRB1
Genomic Map