Assay Details
Gene Symbol: TARBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015151872.2 | XP_015007358.2 | 810 | 92 | ||
XM_015151873.2 | XP_015007359.2 | 408 | 92 | |||
XM_015151875.2 | XP_015007361.2 | 668 | 92 | |||
XM_028829570.1 | XP_028685403.1 | 361 | 92 | |||
GenBank mRNA | JU476973.1 | - | 659 | 92 | ||
JV043368.1 | - | 520 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
