Assay Details
Gene Symbol: STN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266590.1 | NP_001253519.1 | 1072 | 62 | ||
XM_028853531.1 | XP_028709364.1 | 1504 | 62 | |||
GenBank mRNA | JU322960.1 | - | 1101 | 62 | ||
JU477278.1 | - | 992 | 62 | |||
JV636384.1 | - | 1072 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
