Assay Details
Gene Symbol: B4GALT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257591.1 | NP_001244520.1 | 1227 | 109 | ||
XM_015121716.2 | XP_014977202.1 | 1453 | 109 | |||
XM_028837747.1 | XP_028693580.1 | 1136 | 109 | |||
XM_028837748.1 | XP_028693581.1 | 1237 | 109 | |||
GenBank mRNA | JU337130.1 | - | 1227 | 109 | ||
JU337131.1 | - | 1227 | 109 | |||
JU477869.1 | - | 1110 | 109 | |||
JV567859.1 | - | 1290 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
