Assay Details
Gene Symbol: FKBP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260551.1 | NP_001247480.1 | 983 | 85 | ||
XM_015119659.2 | XP_014975145.1 | 1026 | 85 | |||
GenBank mRNA | JV047858.1 | - | 989 | 85 | ||
JV047859.1 | - | 983 | 85 | |||
JV635579.1 | - | 1226 | 85 | |||
JV658044.1 | - | 1220 | 85 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
