Assay Details
| Gene Symbol: SOX9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032868.1 | NP_001028040.1 | 683 | 101 | ||
| GenBank mRNA | AF322900.1 | - | 683 | 101 | ||
| JV694268.1 | - | 1030 | 101 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SOX9
Genomic Map