Assay Details
| Gene Symbol: STX10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261693.1 | NP_001248622.1 | 301 | 79 | ||
| XM_015122994.1 | XP_014978480.1 | 492 | 79 | |||
| GenBank mRNA | JU336890.1 | - | 296 | 79 | ||
| JU477631.1 | - | 301 | 79 | |||
| JV664368.1 | - | 57 | 79 | |||
More Information
Set Membership: |
|

STX10
Genomic Map