Assay Details
Gene Symbol: UXS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015113053.2 | XP_014968539.2 | 845 | 70 | ||
XM_015113054.2 | XP_014968540.2 | 772 | 70 | |||
XM_015113055.2 | XP_014968541.2 | 686 | 70 | |||
XM_015113056.2 | XP_014968542.2 | 686 | 70 | |||
XM_015113059.2 | XP_014968545.2 | 686 | 70 | |||
XM_015113060.2 | XP_014968546.2 | 686 | 70 | |||
XM_028832269.1 | XP_028688102.1 | 686 | 70 | |||
XM_028832270.1 | XP_028688103.1 | 686 | 70 | |||
XM_028832271.1 | XP_028688104.1 | 223 | 70 | |||
GenBank mRNA | JU322318.1 | - | 655 | 70 | ||
JU477290.1 | - | 679 | 70 | |||
JV045223.1 | - | 665 | 70 | |||
JV048300.1 | - | 680 | 70 | |||
JV662062.1 | - | 653 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
