Assay Details
Gene Symbol: PSMB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257425.1 | NP_001244354.1 | 387 | 59 | ||
GenBank mRNA | JU320972.1 | - | 387 | 59 | ||
JU472901.1 | - | 400 | 59 | |||
JV045803.1 | - | 375 | 59 | |||
JV490744.1 | - | 333 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
