Assay Details
| Gene Symbol: PPP2CA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015140928.2 | XP_014996414.1 | 3367 | 70 | ||
| XM_028849729.1 | XP_028705562.1 | 592 | 70 | |||
| XM_028849730.1 | XP_028705563.1 | 3157 | 70 | |||
| GenBank mRNA | JU319939.1 | - | 817 | 70 | ||
| JU471037.1 | - | 687 | 70 | |||
| JV045156.1 | - | 792 | 70 | |||
| JV703316.1 | - | 822 | 70 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PPP2CA
Genomic Map