Assay Details
| Gene Symbol: RGS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265925.1 | NP_001252854.1 | 295 | 115 | ||
| GenBank mRNA | JU335787.1 | - | 299 | 115 | ||
| JU471433.1 | - | 295 | 115 | |||
| JV637491.1 | - | 301 | 115 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RGS2
Genomic Map