Assay Details
| Gene Symbol: ACOT13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171950.1 | NP_001165421.1 | 290 | 87 | ||
| GenBank mRNA | CK231547.1 | - | 290 | 87 | ||
| JU332310.1 | - | 489 | 87 | |||
| JU471589.1 | - | 398 | 87 | |||
| JV047637.1 | - | 499 | 87 | |||
More Information
Set Membership: |
|

ACOT13
Genomic Map