Assay Details
| Gene Symbol: CTLA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044739.1 | NP_001038204.1 | 666 | 79 | ||
| XM_015110722.2 | XP_014966208.1 | 1795 | 79 | |||
| GenBank mRNA | AF344846.1 | - | 666 | 79 | ||
| JU473564.1 | - | 669 | 79 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CTLA4
Genomic Map