Assay Details
| Gene Symbol: MRPL44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015111171.2 | XP_014966657.1 | 905 | 69 | ||
| XM_028831154.1 | XP_028686987.1 | 905 | 69 | |||
| GenBank mRNA | AY680570.1 | - | 61 | 69 | ||
| JU326925.1 | - | 759 | 69 | |||
| JU476859.1 | - | 876 | 69 | |||
| JV047970.1 | - | 878 | 69 | |||
| JV591199.1 | - | 882 | 69 | |||
| JV659168.1 | - | 895 | 69 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MRPL44
Genomic Map