Assay Details
Gene Symbol: PROS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044726.1 | NP_001038191.1 | 1236 | 84 | ||
XM_015128567.2 | XP_014984053.1 | 1662 | 84 | |||
XM_028843093.1 | XP_028698926.1 | 1272 | 84 | |||
GenBank mRNA | JU326109.1 | - | 1114 | 84 | ||
L31380.1 | - | 1236 | 84 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
