Assay Details
Gene Symbol: LYN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001087049.4 | XP_001087049.2 | 672 | 64 | ||
XM_015145385.2 | XP_015000871.1 | 595 | 64 | |||
GenBank mRNA | JV468636.1 | - | 462 | 64 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
