Assay Details
| Gene Symbol: ELP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194851.1 | NP_001181780.1 | 470 | 86 | ||
| XM_001113386.4 | XP_001113386.1 | 684 | 86 | |||
| XM_015131076.2 | XP_014986562.1 | 562 | 86 | |||
| XM_028843337.1 | XP_028699170.1 | 782 | 86 | |||
| XM_028843338.1 | XP_028699171.1 | 501 | 86 | |||
| GenBank mRNA | JU323417.1 | - | 604 | 86 | ||
| JU473382.1 | - | 616 | 86 | |||
| JV045576.1 | - | 620 | 86 | |||
| JV529782.1 | - | 573 | 86 | |||
More Information
Set Membership: |
|

ELP6
Genomic Map