Assay Details
| Gene Symbol: SLC25A41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001091089.4 | XP_001091089.2 | 1244 | 78 | ||
| XM_028839863.1 | XP_028695696.1 | 846 | 78 | |||
More Information
Set Membership: |
|

SLC25A41
Genomic Map