Assay Details
| Gene Symbol: CER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194495.3 | NP_001181424.1 | 558 | 119 | ||
| GenBank mRNA | JV727114.1 | - | 120 | 119 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CER1
Genomic Map